Anti-IgG2 beta Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: GeneTex
GTX35060
89333-296EA 164.38 USD
89333-296
Anti-IgG2 beta Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Secondary
Antigen: IgG2 beta
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR