Anti-IgG1 Rat Monoclonal Antibody (Biotin)

Supplier: GeneTex
GTX35053
89262-118EA 212.09 USD
89262-118
Anti-IgG1 Rat Monoclonal Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: IgG1
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR