Anti-IgG1 Rat Monoclonal Antibody

Supplier: GeneTex
GTX35051
89292-546EA 121.14 USD
89292-546
Anti-IgG1 Rat Monoclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR