Anti-IgG Rat Polyclonal Antibody

Supplier: GeneTex
GTX35047
89336-040EA 131.18 USD
89336-040
Anti-IgG Rat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR