Anti-IgM Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX35038
89262-114EA 97.98 USD
89262-114
Anti-IgM Rabbit Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR