Anti-IgM Mouse Monoclonal Antibody (PE (Phycoerythrin))

Supplier: GeneTex
GTX35034
89265-368EA 185.62 USD
89265-368
Anti-IgM Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Secondary
Antigen: IgM
Clonality: Monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR