Anti-IgG3 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Supplier: GeneTex
GTX35030
89265-366EA 155.53 USD
89265-366
Anti-IgG3 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Secondary
Antigen: IgG3
Clonality: Monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR