Anti-IgG3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: GeneTex
GTX35028
89281-792EA 156.43 USD
89281-792
Anti-IgG3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Secondary
Antigen: IgG3
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR