Anti-SYN1 Rabbit Polyclonal Antibody

GTX31106
89303-494EA 497.32 USD
89303-494
Anti-SYN1 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Synapsin I (phospho Ser549)

Recommended Starting Dilutions:Western Blotting: Max Dilution 1/1000Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: SYN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR