Anti-NPY4R Sheep Polyclonal Antibody

Supplier: GeneTex
GTX31074
89334-130EA 482.66 USD
89334-130
Anti-NPY4R Sheep Polyclonal Antibody
Antibodies
Sheep polyclonal antibody to NPY4R

Recommended Starting Dilutions:Immunohistology - Paraffin: 1/50 - 1/500Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: NPY4R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR