Anti-NPY1R Sheep Polyclonal Antibody

Supplier: GeneTex
GTX31073
89334-128EA 660.25 USD
89334-128
Anti-NPY1R Sheep Polyclonal Antibody
Antibodies
Sheep polyclonal antibody to NPY1R

Recommended Starting Dilutions:Western Blotting: 1/100 - 1/1000Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: NPY1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR