Anti-Neuropeptide Q/NPQ Rabbit Polyclonal Antibody

GTX31070
89298-420EA 401.7 USD
89298-420
Anti-Neuropeptide Q/NPQ Rabbit Polyclonal Antibody
Type: Primary
Antigen: Neuropeptide Q/NPQ
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR