Anti-GRM3 Rabbit Polyclonal Antibody

GTX31058
89303-472EA 428.57 USD
89303-472
Anti-GRM3 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Metabotropic Glutamate receptor 3

We recommend the following starting dilutions: immunohistochemistry 3 ug/ml to 17 ug/ml, Western Blot 3 ug/ml to 17 ug/ml, Immunocytochemistry 3 ug/ml to 17 ug/ml, Immunohistochemistry-Paraffin 3 ug/ml to 17 ug/ml, Immunohistochemistry-Frozen 3 ug/ml to 17 ug/ml. Optimal concentration should be determined experimentally by the end user.

Type: Primary
Antigen: GRM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR