Anti-LDOPA Rabbit Polyclonal Antibody

GTX31051
89303-490EA 514.4 USD
89303-490
Anti-LDOPA Rabbit Polyclonal Antibody
Type: Primary
Antigen: LDOPA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR