Anti-HTT Mouse Monoclonal Antibody [clone: HDA3E10]

GTX31044
89303-484EA 531.42 USD
89303-484
Anti-HTT Mouse Monoclonal Antibody [clone: HDA3E10]
Mouse monoclonal antibody to Huntingtin

We recommend the following starting dilutions: immunofluorescence, Immunohistochemistry-Frozen, immunoprecipitation, Western Blot. Optimal concentration should be determined experimentally by the end user.

Type: Primary
Antigen: HTT
Clonality: Monoclonal
Clone: HDA3E10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR