Anti-HRH3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX31041
89294-686EA 609.89 USD
89294-686
Anti-HRH3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Histamine receptor H3

We recommend the following starting dilutions: ELISA 1:1000-1:5000, Western Blot 1:500-1:1000,. Optimal concentration should be determined experimentally by the end user.

Type: Primary
Antigen: HRH3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR