Anti-HRH4 Rabbit Polyclonal Antibody

GTX31040
89303-466EA 428.57 USD
89303-466
Anti-HRH4 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Histamine H4 receptor

We recommend the following starting dilutions: Immunohistochemistry-Paraffin 2-12 ug/ml. Optimal concentration should be determined experimentally by the end user.

Type: Primary
Antigen: HRH4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR