Anti-THOP1 Mouse Monoclonal Antibody [clone: 4D6]

Supplier: GeneTex
GTX30631
89264-888EA 554.7 USD
89264-888
Anti-THOP1 Mouse Monoclonal Antibody [clone: 4D6]
Antibodies
Mouse monoclonal antibody to Thimet Oligopeptidase [4D6 ]

This antibody may be used in Western blot where a band is seen at ~80 kDa, representing the THOP1 protein. Past productions of this antibody have also been used for immunocytochemistry (human, monkey, and rat brain and testis formalin-fixed paraffin-embedded sections) but this lot has not yet been tested in such an application.Suggested working dilutions: *Western blot - 1:750-1:3,000Immunocytochemistry - (+)*The investigator should determine the optimal working dilution for a specific application.

Type: Primary
Antigen: THOP1
Clonality: Monoclonal
Clone: 4D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Monkey, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR