Anti-ARFGAP1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX21308
89286-196EA 395.68 USD
89286-196
Anti-ARFGAP1 Goat Polyclonal Antibody
Antibodies
Western Blot - Approx 55-60kDa band seen in Mouse Brain extracts. Recommended for use at 1.0-3.0?g/ml.

Type: Primary
Antigen: ARFGAP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR