Anti-CD15 Mouse Monoclonal Antibody [clone: MEM-158]

Supplier: GeneTex
GTX20665
89337-166EA 661.39 USD
89337-166
Anti-CD15 Mouse Monoclonal Antibody [clone: MEM-158]
Antibodies
FACS: Use at an assay dependant concentration. IP: Use at an assay dependant concentration. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: MEM-158
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR