Anti-RBX1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX15517
89317-088EA 376.52 USD
89317-088
Anti-RBX1 Rabbit Polyclonal Antibody
Antibodies
IHC-P: Ready to use for 10 min at room temperature. Staining of formalin fixed tissues requires boiling tissue sections in 10mMcitrate buffer, pH 6.0 for 10 min followed by cooling at room temperature for 20 min. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: RBX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR