Anti-WNT1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX15252
89317-064EA 376.52 USD
89317-064
Anti-WNT1 Rabbit Polyclonal Antibody
Antibodies
IHC-P: Use neat for 10 min at RT. Boil tissue sections in 10mM citrate buffer, pH 6.0 for 10 min followed by cooling at RT for 20 min. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: WNT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR