Anti-OCT2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX15115
89285-864EA 391.69 USD
89285-864
Anti-OCT2 Rabbit Polyclonal Antibody
Antibodies
IHC-P: Use at an assay dependent dilution. Perform heat mediated antigen retrieval before commencing with IHC staining protocol. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: OCT2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR