Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GYPA Mouse Monoclonal Antibody [clone: HIR2]

Supplier: Genetex

GTX15009
89317-346
Anti-GYPA Mouse Monoclonal Antibody [clone: HIR2]
FACS: Tested by flow cytometric analysis of human peripheral blood leukocytes and cell agglutination assay. Can be used at approximately 0.1 mg per million cells. IHC-Fr: Use at an assay dependent dilution. Agglutination studies: The antibody agglutinates untreated RBCs but it fails to agglutinate papain-treated cells. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: GYPA
Clonality: Monoclonal
Clone: HIR2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty