Anti-INK4c Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX14974
89328-260EA 658.42 USD
89328-260
Anti-INK4c Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: INK4c
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR