Anti-CPB2 Mouse Monoclonal Antibody [clone: 16C5]

Supplier: GeneTex
GTX14771
89336-858EA 341.13 USD
89336-858
Anti-CPB2 Mouse Monoclonal Antibody [clone: 16C5]
Antibodies
ELISA: Use at an assay dependent dilution. EIA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 50.8 kDa. Can be paired for ELISA with this antibody as capture with GTX14756 Mouse monoclonal [13D5] to Carboxypeptidase B2 as detection. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CPB2
Clonality: Monoclonal
Clone: 16C5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR