Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CPB2 Mouse Monoclonal Antibody [clone: 1900]

Supplier: Genetex

GTX14767
89336-856
Anti-CPB2 Mouse Monoclonal Antibody [clone: 1900]
ELISA: Use at an assay dependent dilution. EIA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 50.8 kDa. Can be paired for ELISA with this antibody as detection with GTX14756 Mouse monoclonal [13D5] to Carboxypeptidase B2 or GTX14763 Mouse monoclonal [15A10] to Carboxypeptidase B2 as capture. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CPB2
Clonality: Monoclonal
Clone: 1900
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty