Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FAF2 Goat Polyclonal Antibody

Supplier: Genetex

GTX14759
89287-922
Anti-FAF2 Goat Polyclonal Antibody
Approx 50-55kDa band observed in Human Testis, MOLT-4 and Jurkat lysates (predicted MW of 51kDa according to NP_055428). Recommended for use at 1-3µg/ml.

Type: Primary
Antigen: FAF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty