Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BS69 Goat Polyclonal Antibody

Supplier: Genetex

GTX14747
89281-756EA 370.6 USD InStock
89281-756
Anti-BS69 Goat Polyclonal Antibody
Antibodies
Preliminary experiments gave no signal but low background in human heart and brain lysates at up to 1µg/ml. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates? This antibody is expected to recognise both reported Human isoforms. Variants (NP_006615.2; NP_001154954.1) encode the same isoform.

Type: Primary
Antigen: BS69
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty