Anti-Phantom Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX14675
89261-706EA 585.03 USD
89261-706
Anti-Phantom Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Phantom
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR