Anti-TNNI3 Mouse Monoclonal Antibody [clone: 267]

Supplier: GeneTex
GTX14530
89336-848EA 659.34 USD
89336-848
Anti-TNNI3 Mouse Monoclonal Antibody [clone: 267]
Antibodies
Recommended pairs for ELISA: M18 (GTX10236) + 560 (GTX38795) mixture for capture and 19C7 (GTX28289) + 267 (this antibody) mixture for detection MF4 (GTX10239) + 560 (GTX38795) mixture for capture and 19C7 (GTX28289) + 267 (this antibody) mixture for detection

Type: Primary
Antigen: TNNI3
Clonality: Monoclonal
Clone: 267
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR