Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EME1 Mouse Monoclonal Antibody [clone: mta317h2/1]

Supplier: Genetex

GTX14392
89328-246EA 358.96 USD InStock
89328-246
Anti-EME1 Mouse Monoclonal Antibody [clone: mta317h2/1]
Antibodies
WB: Use at an assay dependent dilution. Predicted molecular weight: 68 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: EME1
Clonality: Monoclonal
Clone: mta317h2/1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty