Anti-ENSA Chicken Polyclonal Antibody

Supplier: GeneTex
GTX14297
89261-656EA 692.95 USD
89261-656
Anti-ENSA Chicken Polyclonal Antibody
Antibodies
For WB: Use at an assay dependent dilution. Predicted molecular weight: 15 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: ENSA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR