Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EIF4EBP2 (eukaryotic translation initiation Factor 4E binding Protein 2) Chicken Polyclonal Antibody

Supplier: Genetex

GTX14295
89261-654
Anti-EIF4EBP2 (eukaryotic translation initiation Factor 4E binding Protein 2) Chicken Polyclonal Antibody
For WB: Use at an assay dependent dilution. Predicted molecular weight: 14 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: EIF4EBP2 (eukaryotic translation initiation Factor 4E binding Protein 2)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty