Anti-FABP4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX13979
89278-872EA 602.26 USD
89278-872
Anti-FABP4 Rabbit Polyclonal Antibody
Antibodies
ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 15 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: FABP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR