Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PS2 Mouse Monoclonal Antibody [clone: 198C679.2.1]

Supplier: Genetex

GTX13926
89280-122
Anti-PS2 Mouse Monoclonal Antibody [clone: 198C679.2.1]
WB: Use at a dilution of 1/250 - 1/500. Detects a band of approximately 55 kDa (predicted molecular weight: 54 kDa). It tested negative in mouse NIH 3T3 cells. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PS2
Clonality: Monoclonal
Clone: 198C679.2.1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty