Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL21R Rabbit Polyclonal Antibody

Supplier: Genetex

GTX13856
89280-102
Anti-IL21R Rabbit Polyclonal Antibody
For ELISA: Use at a concentration of 0.5 - 2 μg/ml. For WB: Use at a concentration of 0.5 - 2 μg/ml. Detects a band of approximately 60 kDa. The 60 kD IL21 Receptor band is detected in 30 ug/lane of whole Raji cell lysate. Bands at 45 kD and 35 kD are also seen. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: IL21R
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty