Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CHFR Goat Polyclonal Antibody

Supplier: Genetex

GTX13773
89278-838EA 421.12 USD InStock
89278-838
Anti-CHFR Goat Polyclonal Antibody
Antibodies
Preliminary experiments gave an approx 80kDa band in human brain lysates at 0.5ug/ml, this band was successfully blocked by incubation with the immunising peptide. Please note that currently we cannot find an explanation in the literature for the band we observe given the predicted size of approx. 72kDda according to NP_060693. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported? In paraffin embedded human brain (cerebellum) shows stained Purkinje cells. Recommended concentration, 2.5µg/ml. This antibody is expected to recognise all reported isoforms.

Type: Primary
Antigen: CHFR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty