Anti-DBC2 (Rho-related BTB Domain containing 2) Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX13738
89278-836EA 602.26 USD
89278-836
Anti-DBC2 (Rho-related BTB Domain containing 2) Rabbit Polyclonal Antibody
Antibodies
WB: Use at a dilution of 1/200. Predicted molecular weight: 83 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: DBC2 (Rho-related BTB Domain containing 2)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR