Anti-ASB3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX13715
89278-834EA 602.26 USD
89278-834
Anti-ASB3 Rabbit Polyclonal Antibody
Antibodies
WB: Use at a dilution of 1/500 - 1/1000. Predicted molecular weight: 62 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ASB3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR