Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CLLD8 / SETDB2 (SET Domain bifurcated 2) Goat Polyclonal Antibody

Supplier: Genetex

GTX13712
89287-892EA 388.49 USD InStock
89287-892
Anti-CLLD8 / SETDB2 (SET Domain bifurcated 2) Goat Polyclonal Antibody
Antibodies
Approx 80kDa band observed in human heart lysates (calculated MW of 81.9kDa according to NP_114121.1). Recommended concentration: 0.3-1µg/ml. This product is not suitable in WB on Mouse Heart and Rat Heart lysates. This antibody is expected to recognize both reported isoforms (NP_114121.2; NP_001153780.1).

Type: Primary
Antigen: CLLD8 / SETDB2 (SET Domain bifurcated 2)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty