Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CGBP (CXXC finger Protein 1) Rabbit Polyclonal Antibody

Supplier: Genetex

GTX13695
89278-826EA 421.12 USD InStock
89278-826
Anti-CGBP (CXXC finger Protein 1) Rabbit Polyclonal Antibody
Antibodies
WB: Use at a dilution of 1/1000. Detects a band of approximately 88 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CGBP (CXXC finger Protein 1)
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty