Anti-PDCD7 Mouse Monoclonal Antibody [clone: 46C713]

Supplier: GeneTex
GTX13650
89261-522EA 614.99 USD
89261-522
Anti-PDCD7 Mouse Monoclonal Antibody [clone: 46C713]
Antibodies
WB: Use at a concentration of 1 - 2 ?g/ml. Predicted molecular weight: 54 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PDCD7
Clonality: Monoclonal
Clone: 46C713
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR