Anti-ARL6IP5 Goat Polyclonal Antibody

Supplier: GeneTex
GTX13618
89287-888EA 607.87 USD
89287-888
Anti-ARL6IP5 Goat Polyclonal Antibody
Antibodies
Approx 20kDa band observed in Human Brain lysates (predicted MW of 21.6kDa according to NP_006398.1). Recommended for use at 0.1-0.3µg/ml.

Type: Primary
Antigen: ARL6IP5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR