Anti-FLOT1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX13493
89261-516EA 614.81 USD
89261-516
Anti-FLOT1 Goat Polyclonal Antibody
Antibodies
Approx 48kDa band observed in lysates of cell line H460 (calculated MW of 47.4kDa according to NP_005794.1). In transfected HEK293 transiently expressing FLOT1 a band of approx. 48kDa is observed. This band is not observed in the non-transfected HEK293. Recommended for use at 0.3-1µg/ml.

Type: Primary
Antigen: FLOT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR