Anti-HIV1 GP120 Mouse Monoclonal Antibody [clone: HIV1gp120]

Supplier: GeneTex
GTX13411
89336-840EA 659.34 USD
89336-840
Anti-HIV1 GP120 Mouse Monoclonal Antibody [clone: HIV1gp120]
Antibodies
For ELISA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: HIV1 GP120
Clonality: Monoclonal
Clone: HIV1gp120
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR