Anti-GHRHR Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX13342
89277-770EA 481.52 USD
89277-770
Anti-GHRHR Rabbit Polyclonal Antibody
Antibodies
IHC-P: Use at a dilution of 1/100 - 1/200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: GHRHR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR