Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GALR1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX13338
89277-768EA 402.27 USD InStock
89277-768
Anti-GALR1 Rabbit Polyclonal Antibody
Antibodies
IHC-P: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: GALR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty