Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SYN2 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX13258
89317-910
Anti-SYN2 Rabbit Polyclonal Antibody
WB: Use at a dilution of 1/5000. This dilution was sufficient for detection of Synapsin II in 15 μg of rat brain tissue extract. Detects bands of approximately 74 and 54 kDa, corresponding to the apparent molecular masses of synapsin IIa and IIb.(predicted molecular weight: 57.5 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SYN2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty