Anti-GPRC1B Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX13192
89277-738EA 616.87 USD
89277-738
Anti-GPRC1B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GPRC1B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR